missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10784-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.Specifications
ASF1A | |
Polyclonal | |
Unconjugated | |
ASF1A | |
ASF1A, CCG1 interacting factor A, CGI 98, CIA, hAsf1, hAsf1a, hCIA, Histone chaperone ASF1A, HSPC146 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25842, 294408, 66403 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CQE6, Q9Y294 | |
ASF1A | |
ASF1A Fusion Protein Ag1233 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction