Learn More
Thermo Scientific Chemicals Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Product Code. p-5384531
0.5 mg, 0.50mg
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.50mg
1mg
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Common Name Flag PeptideĂ– Sigma-Aldrich
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 1013.00
Product Type Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence DYKDDDDK Peptide
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys >