missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
4652.90 SEK
Specifications
Antigen | ASPM |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884735
|
Proteintech
26223-1-AP-20UL |
20 ÎĽL |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874735
|
Proteintech
26223-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is the human ortholog of the Drosophila melanogaster ′abnormal spindle′ gene (asp), which is essential for normal mitotic spindle function in embryonic neuroblasts. Studies in mouse also suggest a role of this gene in mitotic spindle regulation, with a preferential role in regulating neurogenesis. Mutations in this gene are associated with microcephaly primary type 5. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
ASPM | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8IZT6 | |
259266 | |
ASPM Fusion Protein Ag24166 | |
Primary | |
-20°C | |
ASPM |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASP, Asp homolog, ASPM, Calmbp1, MCPH5 | |
ASPM | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASPM Rabbit anti-Human, Polyclonal, Proteintech