missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASS1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D1A11, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ASS1 |
---|---|
Clone | 1D1A11 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887545
|
Proteintech
66036-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877545
|
Proteintech
66036-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.Specifications
ASS1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Argininosuccinate synthase, argininosuccinate synthetase 1, ASS, ASS1, Citrulline aspartate ligase, CTLN1 | |
ASS1 | |
IgG1 | |
Protein G | |
Antibody |
1D1A11 | |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Pig, Rat, Mouse | |
P00966, P09034, P16460 | |
11898, 25698, 414411, 445 | |
ASS1 Fusion Protein Ag9314 | |
Primary | |
-20°C | |
ASS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASS1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D1A11, Proteintech