Learn More
Hecht Karl™ Assistent™ Automatic Burette
594.00 SEK - 694.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10237671
|
Hecht Karl™
40820010 |
10 mL |
594.00 SEK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10630012
|
Hecht Karl™
40820005 |
5 mL |
694.00 SEK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Specifications
Glass | |
Automatic Buret |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hecht Karl™ Assistent™ Automatic Burette