missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11264-1-AP-20UL
This item is not returnable.
View return policy
Description
ATG12, a member of the autophagy protein family, forms a conjugate with ATG5 this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1.Specifications
ATG12 | |
Polyclonal | |
Unconjugated | |
ATG12 | |
APG12, APG12 like, APG12L, ATG12, Autophagy related protein 12, FBR93, HAPG12, Ubiquitin like protein ATG12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361321, 67526, 9140 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94817, Q2TBJ5, Q9CQY1 | |
Atg12 | |
ATG12 Fusion Protein Ag1791 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATG12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction