missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11262-2-AP-20UL
This item is not returnable.
View return policy
Description
ATG3 (APG3) is a widely expressed conjugating enzyme for APG8 lipidation, an essential step for the initiation of autophagy. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. Autophagy is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG3 functions as an E2-like enzyme during the initial stages of autophagosome formation by catalyzing the formation of the Atg8-phosphatidylethanolamine (Atg8-PE) conjugate, which is critical for autophagy.Specifications
ATG3 | |
Polyclonal | |
Unconjugated | |
ATG3 | |
APG3, APG3 LIKE, APG3L, ATG3, Autophagy related protein 3, hApg3, PC3 96, Protein PC3 96 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171415, 64422, 67841 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AZ50, Q9CPX6, Q9NT62 | |
Atg3 | |
ATG3 Fusion Protein Ag1785 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction