missing translation for 'onlineSavingsMsg'
Learn More

ATG4D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16924-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886694

  • 3665.00 SEK / 150µL
Estimated Shipment: 28-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases.
Specifications

Specifications

ATG4D
Polyclonal
Unconjugated
ATG4D
APG4 D, APG4D, ATG4D, AUTL4, Autophagin 4, Cysteine protease ATG4D
Rabbit
Antigen Affinity Chromatography
RUO
235040, 686505, 84971
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86TL0, Q8BGV9
ATG4D
ATG4D Fusion Protein Ag10307
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers