Learn More
ATOX1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22641-1-AP-150UL
This item is not returnable.
View return policy
Description
The ATOX1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ATOX1. This antibody recognizes human antigen. The ATOX1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ATOX1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
475 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
340 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATOX1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL
Spot an opportunity for improvement?Share a Content Correction