Learn More
ATOX1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22641-1-AP-150UL
 View more versions of this product
Product Code. 16882015
  • 4652.90 SEK / 150µL
Description
The ATOX1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ATOX1. This antibody recognizes human antigen. The ATOX1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ATOX1
Polyclonal
Unconjugated
150 μL
475
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
340 μg/mL
Rabbit
Primary
Human
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ATOX1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL