missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP8A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21565-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
ATP8A1 | |
Polyclonal | |
Unconjugated | |
ATP8A1 | |
ATP8A1, ATPase II, ATPASEII, ATPIA, ATPP2, Chromaffin granule ATPase II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10396, 11980, 289615 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70704, Q9Y2Q0 | |
Atp8a1 | |
ATP8A1 Fusion Protein Ag16071 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |