missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP8A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ATP8A1 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891155
|
Proteintech
21565-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881155
|
Proteintech
21565-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
ATP8A1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P70704, Q9Y2Q0 | |
10396, 11980, 289615 | |
ATP8A1 Fusion Protein Ag16071 | |
Primary | |
-20°C | |
ATP8A1 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ATP8A1, ATPase II, ATPASEII, ATPIA, ATPP2, Chromaffin granule ATPase II | |
Atp8a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATP8A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech