missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ATPAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15797-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 beta subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. Alternatively spliced transcript variants have been identified, but the biological validity of some of these variants has not been determined.Specifications
ATPAF1 | |
Polyclonal | |
Unconjugated | |
ATPAF1 | |
ATP11 homolog, ATP11p, ATPAF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230649, 313510, 64756 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5TC12, Q811I0 | |
ATPAF1 | |
ATPAF1 Fusion Protein Ag8506 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |