Learn More
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67057-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889455
  • 1656.73 SEK / 20µL
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
The ATX3, ATXN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATX3, ATXN3. This antibody recognizes human, mouse, pig antigen. The ATX3, ATXN3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ATX3 ATXN3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig, Mouse
Western Blot, ELISA
1F7E10
Unconjugated
20 μL
4287
IgG2a