Learn More
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67057-1-IG-150UL
Description
The ATX3, ATXN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATX3, ATXN3. This antibody recognizes human, mouse, pig antigen. The ATX3, ATXN3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ATX3 ATXN3 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Pig, Mouse |
Western Blot, ELISA | |
1F7E10 | |
Unconjugated | |
150 ÎĽL | |
4287 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.