missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13726-1-AP-20UL
This item is not returnable.
View return policy
Description
AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain thay may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precusor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.Specifications
AUP1 | |
Polyclonal | |
Unconjugated | |
AUP1 | |
ancient ubiquitous protein 1, AUP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11993, 550, 680423 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1L134, P70295, Q9Y679 | |
AUP1 | |
AUP1 Fusion Protein Ag4680 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AUP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction