Learn More
Bollé Safety AXIS Safety Glasses, Polycarbonate Frame, Polycarbonate Lens
Click to view available options
Lens Tint:
Clear
Contrast
Unit Size:
Each
Description
- Upper protection
- Pivoting temples
- Non-slip TIPGRIP temples
- Adjustable non-slip bridge
- Anti-static
Specifications
Specifications
Certifications/Compliance | European standard |
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Safety glasses; Axis |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
Documents
Product Information
Specification Sheets
AXIS
File size: 744 kB
Product Content Correction
Product Title
Bollé Safety AXIS Safety Glasses, Polycarbonate Frame, Polycarbonate Lens >
Spot an opportunity for improvement?Share a Content Correction