missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AXUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18162-1-AP-150UL
This item is not returnable.
View return policy
Description
Axin, an important regulator of beta-catenin, is frequently mutated in human hepatocellular carcinomas (HCCs), and transduction of the wild-type Axin gene (AXIN1) induces apoptosis in HCC cells as well as in colon cancer cells. A novel gene, AXUD1 (AXIN1 up-regulated), has enhanced expression in response to exogenously expressed AXIN1. It is expressed in all human tissues examined, most abundantly in lung, placenta, skeletal muscle, pancreas and leukocytes. Data suggests that AXUD1 may have a tumor-suppressor function in those organs.Specifications
AXUD1 | |
Polyclonal | |
Unconjugated | |
CSRNP1 | |
AXIN1 up regulated 1, AXUD1, CSRNP1, CSRNP1 Ens, FAM130B, TAIP 3, URAX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215418, 363165, 64651 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59054, Q96S65 | |
Csrnp1 | |
AXUD1 Fusion Protein Ag6158 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AXUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction