Learn More
Axygen™ FDSS™ Robotic Tips
For Hamamatsu FDSS series systems
Brand:  Axygen™ FDSS-384-BK-R
Additional Details : Weight : 10.10000kg
 View more versions of this product
Product Code. 12702846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.
Specifications
Specifications
50 μL
Autoclavable
Rack
Hamamatsu FDSS 6000 System
Black
DNase, RNase, and Pyrogen free
Automation