Learn More
Corning™ Axygen™ Polypropylene Universal Pipette Tip For Maximum Recovery Compat Pipettes
Brand: Corning™ TE-204-Y-L
This item is not returnable.
View return policy
Description
Order Info
CapacityMetric
Specifications
200 ÎĽL | |
Yellow | |
For Ultra-low Retention and High Yield | |
Universal | |
Yes | |
Polypropylene |
Non-filtered | |
DNase, RNase Free | |
Hinged Rack | |
Maximum Recovery Compatible Pipette | |
Axygen |
Product Content Correction
Product Title
Corning™ Axygen™ Polypropylene Universal Pipette Tip For Maximum Recovery Compat Pipettes > Volume: 200 μL
Spot an opportunity for improvement?Share a Content Correction