missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALTL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14601-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a beta-1, 3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).Specifications
B3GALTL | |
Polyclonal | |
Unconjugated | |
B3GLCT | |
B3GALTL, B3Glc T, B3GLCT, B3GTL, Beta 1,3 glucosyltransferase, beta3Glc T, Gal T | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
145173, 381694 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6Y288, Q8BHT6 | |
B3glct | |
B3GALTL Fusion Protein Ag6146 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
B3GALTL Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction