missing translation for 'onlineSavingsMsg'
Learn More

B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10535-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886973

  • 3665.00 SEK / 150µL
Estimated Shipment: 27-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
Specifications

Specifications

B4GALT7
Polyclonal
Unconjugated
B4GALT7
B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1
Rabbit
Antigen Affinity Chromatography
RUO
11285
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UBV7
B4GALT7
B4GALT7 Fusion Protein Ag0793
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers