Learn More
Product Code | Brand | For Use With (Equipment) | Type | Filter Size | Resin Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Filter Size | Resin Type | Price | Quantity & Availability | |||
10422962
|
Thermo Scientific™
D502124 |
Easypure II UV, Easypure II UV/UF | Cartridge Pack, Deionized Feed |
6283.73 SEK
Each |
Estimated Shipment: 04-11-2025
Log in to see stock. |
|||||
10895960
|
Thermo Scientific™
D50237 |
Easypure | Mixed Bed COOL Cartridge | 6 in. | Mixed Bed |
931.32 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10728154
|
Thermo Scientific™
D502125 |
Easypure II UV, Easypure II UV/UF | Cartridge Pack, RO or Distilled Feed |
6283.73 SEK
Each |
In Stock
Log in to see stock. |
|||||
10041672
|
Thermo Scientific™
D502126 |
Easypure II, Easypure UF | Cartridge Pack, Deionized Feed |
6106.22 SEK
Each |
Estimated Shipment: 29-10-2025
Log in to see stock. |
|||||
10133502
|
Thermo Scientific™
D502127 |
Easypure II, Easypure UF | Cartridge Pack, RO or Distilled Feed |
5774.87 SEK
Each |
Estimated Shipment: 22-12-2025
Log in to see stock. |
|||||
10498453
|
Thermo Scientific™
FL1192X1 |
Easypure UF, Easypure II UV/UF | Ultrafilter |
13490.49 SEK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
11426348
|
Thermo Scientific™
D50230 |
Easypure II | Cartridge and Filter | 2/3 Macroreticular Resin |
1917.07 SEK
Each |
In Stock
Log in to see stock. |
||||
10502074
|
Thermo Scientific™
D50245 |
EasyPure II RF | Sanitation Cartridge |
1166.81 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
- Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
- Type 1 filter
D502124:
- Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
D502127:
- Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
- Type 1 filter
D502125:
- Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters