Learn More
Thermo Scientific™ Barnstead™ Final Filter
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems.
Brand: Thermo Scientific™ D3750
1439.65 SEK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems. Compatible with Thermo Scientific™ B-Pure™, Nanopure™, Easypure™ II, Easypure™ II RF, Easypure™ RoDi, E-Pure™ (3,4 holder) and TII systems.
Specifications
0.2µm Final Filter | |
0.2 μm | |
B-Pure, Nanopure, Easypure II, Easypure II RF, Easypure RoDi, E-Pure (3,4 holder), and TII systems | |
Final Filter |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now