missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | BBS7 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881684
|
Proteintech
13458-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871684
|
Proteintech
13458-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
BBS7 | |
Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q8IWZ6, Q8K2G4 | |
361930, 55212, 71492 | |
BBS7 Fusion Protein Ag4314 | |
Primary | |
-20°C | |
BBS7 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Bardet Biedl syndrome 7, BBS2 like protein 1, BBS2L1, BBS7 | |
Bbs7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BBS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech