Learn More
BBS8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12505-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The BBS8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS8. This antibody recognizes human, mouse, rat antigen. The BBS8 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
BBS8 | |
Polyclonal | |
Unconjugated | |
150 μL | |
123016 | |
IgG |
Western Blot, Immunoprecipitation, ELISA | |
353 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |