Learn More
BCL11B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55414-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to BCL11B. The calculated molecular weight of BCL11B is 95 kDa, but modified BCL11B protein is about 116 kDa.
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell maligcies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.Specifications
BCL11B | |
Polyclonal | |
Unconjugated | |
BCL11B | |
B cell CLL/lymphoma 11B, B cell lymphoma/leukemia 11B, BCL11B, COUP TF interacting protein 2, CTIP 2, CTIP2, hRit1, hRIT1 alpha, RIT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58208, 64919 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99PV8, Q9C0K0 | |
BCL11B | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
BCL11B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction