Learn More
Abnova™ BCL2 Split FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
Content And Storage | Store at 4°C in the dark |
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | B-cell CLL/lymphoma 2 |
Gene Alias | Bcl-2 |
Gene Symbol | BCL2 |
Gene ID (Entrez) | 596 |
Product Type | DNA/RNA |
Quantity | 200 μL |
Form | Liquid |
Product Content Correction
Product Title
Abnova™ BCL2 Split FISH Probe >
Spot an opportunity for improvement?Share a Content Correction