Learn More
BD Mouse Th1/Th2/Th17 Phenotyping Kit
Simultaneously view Th17, Th2 and Th17 cells in mouse model systems
Brand:  BD 560758
9205.21 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15840469
  • 9210.00 SEK / Each
Includes: Mouse Th1/Th2/Th17 Phenotyping Cocktails (CD4 PerCP-Cy™5.5, IL-17A PE, IFN- γFITC, IL-4 APC), BD Cytofix™ Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation
  • Ensures indentification and characterization of the nature of the T helper cell types
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BD Mouse Th1/Th2/Th17 Phenotyping Kit > 1 Kit (50 Tests)
For research use only. Not for diagnostic or therapeutic procedures.