Learn More
Bel-Art™ SP Scienceware™ Modular Ultra-Low Five-Box Freezer Racks
Link together side-by-side or can be securely stacked for horizontal and vertical storage.
Brand: Bel-Art™ H18992-0002
Additional Details : Weight : 1.19050kg
Description
- Configured to customize freezer space
- Includes pull out drawer for easy sample retrieval even from back of freezer
- Each lightweight, wire formed rack holds 5 freezer boxes up to 5.25 x 5.25 x 2 in. H
- Available in 4 colors for easy sample identification
- Top cover provides flat shelf space for additional storage
Specifications
Ultra Low Freezer Rack | |
White | |
Ultra low freezer racks | |
68.6 x 15.2 x 8.9cm (27 x 6 x 3.5 in.) |