Learn More
Bel-Art™ SP Scienceware™ Poxygrid 60-Place Test Tube Racks
To organize and process high volumes of samples.
Brand:  Bel-Art™ F18763-0160
Product Code. 13432509
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Steel wire test tube rack
  • Chemical and corrosion-resistant
  • Coated with heat-cured epoxy
  • Unaffected by organic solvents, salts, weak acids and weak alkalis
  • Temperature range: −223°C (−369°F) to 121°C (250°F)
  • Autoclavable
  • With square holes, three decks, slotted grid bottom and reinforced edges
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Poxygrid 60-Place Test Tube Racks > For 18-20mm tubes; Blue; 8.3cm H top deck