missing translation for 'onlineSavingsMsg'
Learn More

SP Scienceware™ Bel-Art™ Sterileware Double Bagged Long Handle Sampling Spoons

Minimizing Contamination of Small Sample Collection

Brand:  SP Scienceware™ H36946-1010

Additional Details : Weight : 0.68040kg

 View more versions of this product

Product Code. 16619755

  • POA / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.

  • Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10-6
  • Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
Specifications

Specifications

Spoon
3 tsp.
Sterile
Certificate of Processing for Sterilization
17.8 cm
4 cm
Long-handled
14.79 mL
White
7 in.
1 9/16 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers