Learn More
SP Scienceware™ Bel-Art™ Sterileware Double Bagged Long Handle Sampling Spoons
Minimizing Contamination of Small Sample Collection
Brand:  SP Scienceware™ H36940-1010
Additional Details : Weight : 0.68040kg
 View more versions of this product
Product Code. 16689745
  • 630.00 SEK / Pack of 10
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
  • Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10-6
  • Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
Spoon
0.25 tsp.
Sterile
Certificate of Processing for Sterilization
17.8 cm
2 cm
Long-handled
1.25 mL
White
7 in.
13/16 in.