missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEND4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24711-1-AP-150UL
This item is not returnable.
View return policy
Description
BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.Specifications
BEND4 | |
Polyclonal | |
Unconjugated | |
BEND4 | |
BEN domain containing 4, BEND4, CCDC4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389206, 666938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P86174, Q6ZU67 | |
BEND4 | |
BEND4 Fusion Protein Ag20169 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BEND4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction