Learn More
Thermo Scientific™ BetaSil™ Phenyl HPLC Columns, 3μm Particle Size
High surface area with high bonded phase coverage
Brand: Thermo Scientific™ 70603-102130
Additional Details : Weight : 0.10000kg
Description
- Thermo Scientific chemistry on 100Å silica
Order Info
Other column dimensions including preparative scale are available. Please call Customer Service for more information.
Specifications
2.1 mm | |
3 μm | |
100 Å | |
Phenyl (Ph) | |
11% | |
Yes |
100 mm | |
2 to 9 | |
L11 | |
Silica | |
Reversed Phase | |
Reversed Phase |