Learn More
Thermo Scientific™ BetaSil™ Phenyl HPLC Columns, 3μm Particle Size
High surface area with high bonded phase coverage
5207.60 SEK - 10350.75 SEK
Specifications
Particle Size | 3 μm |
---|---|
pH | 2 to 9 |
Pore Size | 100 Å |
USP Type | L11 |
Stationary Phase | Phenyl (Ph) |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
10108924
|
Thermo Scientific™
70603-052130 |
2.1 mm | 50 mm | Phenyl (Ph) |
10350.75 SEK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
11502991
|
Thermo Scientific™
70603-022130 |
2.1 mm | 20 mm | Phenyl (Ph) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11512991
|
Thermo Scientific™
70603-102130 |
2.1 mm | 100 mm | Phenyl (Ph) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Thermo Scientific chemistry on 100Å silica
Order Info
Other column dimensions including preparative scale are available. Please call Customer Service for more information.
Specifications
3 μm | |
100 Å | |
Phenyl (Ph) | |
11% | |
Yes |
2 to 9 | |
L11 | |
Silica | |
Reversed Phase | |
Reversed Phase |