Learn More
BHLHE40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17895-1-AP-150UL
 View more versions of this product
Product Code. 16887894
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
Specifications
BHLHE40
Polyclonal
Unconjugated
BHLHE40
BHLHB2, BHLHE40, DEC1, SHARP 2, SHARP2, STRA13, Stra14
Rabbit
Antigen Affinity Chromatography
RUO
20893, 79431, 8553
-20°C
Liquid
Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14503, O35185, O35780
BHLHE40
BHLHE40 Fusion Protein Ag12236
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BHLHE40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated