Learn More
Macherey-Nagel™ BioFix™ Lumi Luminous Bacteria Toxicity Test
Freeze-dried, in accordance with DIN EN ISO 11348-3. Macherey-Nagel™ BioFix™ Lumi Luminous Bacteria Toxicity Test are ready-to-use with a shelf life of at least 24 months after production when stored correctly.
4610.00 SEK - 11141.00 SEK
Specifications
Certifications/Compliance | DIN EN ISO 11348-3 |
---|---|
Composition | Vibrio fischeri NRRL B-11177 |
pH Range | 6.0 to 8.5 (Sample) |
Format | Freeze-dried |
For Use With (Application) | Determination of acute toxicity in communal and industrial sewage water, surface water, diluted eluates of soil, waster and abandoned polluted areas |
Product Code | Brand | No. of Tests | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Volume (Metric) | Price | Quantity & Availability | |||||
11340605
|
Macherey-Nagel™
945007 |
200 Tests | 0.2 mL Tubes |
4610.00 SEK
Pack of 10 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
11310605
|
Macherey-Nagel™
|
2000 Tests | 1 mL Tubes | N/A | ||||||
Description
Determination of the acute toxicity on freeze-dried luminescent bacteria in accordance with DIN EN ISO 11348-3. The measuring unit is the natural luminescence of the used microorganism Vibrio fischeri NRRL B-11177. The test system measures the light output of the luminescent bacteria after they have been challenged by a sample and compares it to the light output of a control (reagent blank) that contains no sample. A difference in light output between the sample and the control is attributed to the effect of the sample on the organisms.
Specifications
DIN EN ISO 11348-3 | |
6.0 to 8.5 (Sample) | |
Determination of acute toxicity in communal and industrial sewage water, surface water, diluted eluates of soil, waster and abandoned polluted areas | |
Freeze-dried bacteria: frozen (−20 ±2°C), Flask: refrigerated (2-8°C) | |
Freeze-dried bacteria |
Vibrio fischeri NRRL B-11177 | |
Freeze-dried | |
0 – 100% inhibition | |
Luminescence |
Safety and Handling
missing translation for 'shelfLife' : 24 Months (frozen)
Your input is important to us. Please complete this form to provide feedback related to the content on this product.