Learn More
ACROBiosystems Biotinylated Human CD14 Protein, His, Avitag
Brand: ACROBiosystems CD4-H8228-200ug
Additional Details : Weight : 0.00010kg
Description
Cluster of differentiation 14 (CD14), is a cell surface glycoprotein, and is a is a component of the innate immune system. CD14 is a myelomonocytic differentiation antigen preferentially expressed on monocytes, macrophages, and activated granulocytes. CD14 exists in two forms. Either it is anchored into the membrane by a glycosylphosphatidylinositol tail (mCD14) or it appears in a soluble form (sCD14). Soluble CD14 either appears after shedding of mCD14 (48 kDa) or is directly secreted from intracellular vesicles (56 kDa). CD14 acts as a co-receptor (along with the Toll-like receptor TLR 4 and MD-2) for the detection of bacterial lipopolysaccharide (LPS). CD14 can bind LPS only in the presence of lipopolysaccharide-binding protein (LBP). CD14 has been proposed to be involved in various biological processes, including transportation of other lipids, cell-cell interaction during different immune responses, as well as recognition of apoptotic cells. Although LPS is considered its main ligand, CD14 also recognizes other pathogen-associated molecular patterns. CD14+ cells are monocytes that can differentiate into a host of different cells. CD14 has been shown to interact with Lipopolysaccharide-binding protein.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 38.6 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH10507.1 | |
>95% as determined by SDS-PAGE. | |
Biotinylated Human CD14, His,Avitag (CD4-H8228) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Met 344 (Accession # P08571-1). | |
-20°C | |
CD14 | |
His Tag & Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
Human |
38.6 kDa | |
200 µg | |
Thr 20 - Met 344 | |
CD14 | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |