Learn More
ACROBiosystems Biotinylated Human Integrin alpha 4 beta 7 (ITGA4&ITGB7) Heterodimer Protein, His, Avitag™&Tag Free
Biotinylated Protein

POA
Specifications
Accession Number | NP 000876.3 (ITGA4) and NP_000880.1 (ITGB7) |
---|---|
Molecular Weight (g/mol) | 113.2 kDa (ITGA4) and 81.8 kDa (ITGB7) |
Source | Biotinylated Human ITGA4&ITGB7; Heterodimer Protein, His,Avitag&Tag; Free (IT7-H82W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) (Accession # P13612-1 (ITGA4) and P26010-1 (ITGB7)). |
Immunogen | Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) |
Storage Requirements | -20°C |
Description
Integrin alpha 4 beta 7 is an integrin family adhesion receptor that shares subunits with alpha 4 beta 1 (VLA4) and the E-Cadherin receptor, alpha E beta 7 . Integrin alpha 4 beta 7, which is critical for homing to intestinal mucosa, is induced during T cell activation in Peyer’s patches or mesenteric lymph nodes.Its expression requires signals from local dendritic and stromal cells, including secreted retinoic acid. The HIV-1 envelope protein gp120 binds to the active form of Integrin alpha 4 beta 7, and this may or may not account for the concentration of HIV-1 virus in the gut-associated lymphoid tissue (GALT). Integrin alpha 4 beta 7 may also be involved in lymphocyte trafficking in acute intestinal graft vs. host disease (GVHD).
Biotinylated Human ITGA4&ITGB7 Heterodimer Protein, His,Avitag&Tag Free, produced by co-expression of ITGA4 and ITGB7, has a calculated MW of 113.2 kDa (ITGA4) and 81.8 kDa (ITGB7). Subunit ITGA4 is fused with an acidic tail at the C-terminus and followedSpecifications
NP 000876.3 (ITGA4) and NP_000880.1 (ITGB7) | |
Biotinylated Human ITGA4&ITGB7; Heterodimer Protein, His,Avitag&Tag; Free (IT7-H82W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) (Accession # P13612-1 (ITGA4) and P26010-1 (ITGB7)). | |
-20°C | |
Integrin alpha 4 beta 7 | |
Human | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
113.2 kDa (ITGA4) and 81.8 kDa (ITGB7) | |
Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) | |
Integrin alpha 4 beta 7,ITGA4andITGB7 | |
Precision Avi | |
His Tag and Avi Tag | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.