missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIRC3 Mouse anti-Human, Clone: OTI1A9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800024S

This item is not returnable.
View return policy
Description
BIRC3 Monoclonal antibody specifically detects BIRC3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
BIRC3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13489 | |
Mouse | |
Affinity chromatography | |
RUO | |
330 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
OTI1A9 | |
Unconjugated | |
BIRC3 | |
AIP 2; AIP1; Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW107670; baculoviral IAP repeat containing 3; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 3; Birc2; Birc3; Cellular inhibitor of apoptosis 2; cIAP1; cIAP-1; cIAP2; C-IAP2; cIAP-2; H HIAP1; H MALT2; HAIP1; HIAP1; hIAP-1; HIAP2; IAP homolog; IAP homolog C; IAP1; IAP-1; IAP2; Inhibitor of apoptosis protein 1; MALT2; mammalian IAP homolog C; MIAP1; mIAP-1; MIAP2; MIHB; MIHC; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; RNF49; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1 | |
Human recombinant protein fragment corresponding to amino acids 255-529 of human BIRC3 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BIRC3 Mouse anti-Human, Clone: OTI1A9, TrueMAB™, Origene
Spot an opportunity for improvement?Share a Content Correction