missing translation for 'onlineSavingsMsg'
Learn More
BIS-TRIS propane, 0.2M buffer soln., pH 7.0, Thermo Scientific™ Product Code.: 15435759

BIS-TRIS propane, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Product Code. p-7042729
100 mL, 100mL
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15435759

Brand: Thermo Scientific Alfa Aesar J63555.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material BIS-TRIS propane buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberX31A503 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberU10D515 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberX03F507 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberN26H515 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot NumberF28X879 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BIS-TRIS propane, 0.2M buffer soln., pH 7.0, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.