missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Blank Micro-Filter-Candles

Robu™ Blank Micro-Filter-Candles are highly resistant to most chemicals such as solvents, acids, and alkalis.

Brand:  Robu™ 18012

Additional Details : Weight : 0.02520kg

 View more versions of this product

Product Code. 16882906

  • 1595.00 SEK / Pack of 5

Estimated Shipment: 22-06-2023
to see stock.
Add to basket

Description

Description

  • ISO 4793-80
  • Cylindrical shape
  • Without narrow glass tube
  • One end closed
Specifications

Specifications

Without glass tube
Cylindrical
20 mm
4.5 mm
40 to 100 μm
Micro Filter Candle
9 mm outer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers