missing translation for 'onlineSavingsMsg'
Learn More

BORIS Mouse anti-Human, Mouse, Clone: 2B12E2, Proteintech

Mouse Monoclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen BORIS
Clone 2B12E2
Concentration 0.71 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897045
View Documents
Proteintech
60079-1-IG-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887045
View Documents
Proteintech
60079-1-IG-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
Specifications

Specifications

BORIS
0.71 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BORIS, Cancer/testis antigen 27, CCCTC binding factor, CT27, CTCF like protein, CTCF paralog, CTCF T, CTCFL, Zinc finger protein CTCF T
Ctcfl
IgG1
Caprylic Acid Precipitation
Antibody
2B12E2
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Mouse
Mouse, Human
A2APF3, Q8NI51
140690, 664799
BORIS Fusion Protein Ag1003
Primary
-20°C
CTCFL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BORIS Mouse anti-Human, Mouse, Clone: 2B12E2, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.