missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BORIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11074-2-AP-20UL
This item is not returnable.
View return policy
Description
Polyclonal anti-CFCFL antibody (11074-2-AP) is produced by immunizing animals with part of C-terminal chain of CTCFL and detects a 75 kDa and a 55 kDa band.
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.Specifications
BORIS | |
Polyclonal | |
Unconjugated | |
CTCFL | |
BORIS, Cancer/testis antigen 27, CCCTC binding factor, CT27, CTCF like protein, CTCF paralog, CTCF T, CTCFL, Zinc finger protein CTCF T | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100360757, 140690, 664799 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2APF3, Q8NI51 | |
Ctcfl | |
BORIS Fusion Protein Ag1547 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BORIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction