Learn More
Boron, plasma standard solution, Specpure™ B 1000μg/mL
729.00 SEK - 2645.00 SEK
Specifications
Chemical Name or Material | Boron |
---|---|
Name Note | plasma standard solution, B 1000ÎĽg/mL |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Physical Form | Liquid |
MDL Number | MFCD00134034 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353518
|
Thermo Scientific Alfa Aesar
039147.AD |
50 mL |
729.00 SEK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343518
|
Thermo Scientific Alfa Aesar
039147.AE |
100 mL |
1025.00 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450221
|
Thermo Scientific Alfa Aesar
039147.AP |
500 mL |
2645.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boron | |
23 hPa (17mm Hg) at 20°C | |
MFCD00134034 |
plasma standard solution, B 1000ÎĽg/mL | |
Liquid |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H360FD
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures