Learn More
Boron Solution for ICP, 100mg/L, H2O Matrix, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
Solution Contains | H2O |
Concentration | 100 mg/L |
Quantity | 100 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Product Title
Boron Solution for ICP, 100mg/L, H2O Matrix, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction