Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04030037
 View more versions of this product
Product Code. 10090531
  • 1680.00 SEK / Pack of 2
Only null left
Estimated Shipment: 12-08-2025
Description
  • Cylindrical shape with straight discharge tube
  • With NS socket and PE stopper
  • With solid glass plug
  • With Screwthreade retaining device
  • With scale
Specifications
Specifications
100 mL
Borosilicate Glass
2Pack
Polyethylene
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug > Capacity: 100mL