Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ 04012037
 View more versions of this product
Product Code. 11882642
  • 759.00 SEK / Pack of 2
Only null left
Estimated Shipment: 28-07-2025
Description
  • With NS socket and PE stopper
  • Scale of approx. volume/mL
  • With screwthread retaining device
Specifications
Specifications
100 mL
Borosilicate Glass
2Pack
Polyethylene
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale > Capacity: 100mL