Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
Brand:  Karl Hecht™ 42096096
 View more versions of this product
Product Code. 10083891
  • 1875.00 SEK / Each
Only null left
Estimated Shipment: 30-06-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Duran™ borosilicate glass 3.3.
  • Standard type.
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Fused blue graduation.
  • Thick-walled, solid finish.
  • Clearly legible figures and marks.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
DIN EN ISO 1042
5 L
Standard
1.2 mL
NS Stopper
Polyethylene
Borosilicate Glass
Volumetric Flask
Yes
NS 34/35
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified > Capacity: 5000mL; Stopper Number: NS 34/35; Tolerance: 1.2mL